Learn More
Med Vet International Skin Stapler and Surgical Staple Remover, Sterile, 35 Wide, Each
Catalog No. 502522540
Encompass_Preferred
This skin stapler is affordable and simple. It's simplistic design and quality construction provides consistent and reliable performance. It fits comfortably in your hand to enhance control of skin closure, wound closure and also reduces surgery time. Surgical Staple Remover Included. 35 staples preloaded Individually sterile Lever action staplers are the most popular design on the market Distinctive staple shuttling feature greatly reduces track friction, a primary cause of jamming Short trigger stroke for ease of use and nonstop feed Handle design comfortably fits a wide range of hand sizes Angled head allows visibility of incision to ensure precise staple placement during operation Arrow indicator assists in accurate staple alignment Staple count indicator allows you to view staples remaining to help minimize waste Quality construction and simple design assure reliable consistent performance Regular staples measure 5.4 mm x 3.6 mm; Wide Staples measure 7 mm x 4 mm
Catalog No. 50-252-2540 Supplier Med Vet International Supplier No. 35WS
This skin stapler is affordable and simple. It's simplistic design and quality construction provides consistent and reliable performance. It fits comfortably in your hand to enhance control of skin closure, wound closure and also reduces surgery time. Surgical Staple Remover Included. 35 staples preloaded Individually sterile Lever action staplers are the most popular design on the market Distinctive staple shuttling feature greatly reduces track friction, a primary cause of jamming Short trigger stroke for ease of use and nonstop feed Handle design comfortably fits a wide range of hand sizes Angled head allows visibility of incision to ensure precise staple placement during operation Arrow indicator assists in accurate staple alignment Staple count indicator allows you to view staples remaining to help minimize waste Quality construction and simple design assure reliable consistent performance Regular staples measure 5.4 mm x 3.6 mm; Wide Staples measure 7 mm x 4 mm