Learn More
Med Vet International Skin Stapler, Sterile, 35 Wide, AHS35 Wide, Each
Catalog No. 501180500
Encompass_Preferred
AHS Skin Stapler is a top quality, sterile, single use, skin staple. This stapler comes with (35) wide medical grade stainless steel staples and measure 7.5mm in length, 4.3mm wide, and has a diameter of .6mm. This staper features: A unique ratchet system to prevent jamming Ergonomic, user friendly design Easy to check remaining staples in cartridge Clear view of operating site Short trigger stroke for ease of use and nonstop feed Angled head allows visibility of incision to ensure precise staple placement Alternative to products like ZipStitch instant stitches and Sutures Easy to use For deeper wounds Faster than Stitches Easier to remove than Stitches Fewer issues with scarring
Catalog No. 50-118-0500 Supplier Med Vet International Supplier No. AHS35W
 Restricted Item
Add to Cart
Add to Cart
AHS Skin Stapler is a top quality, sterile, single use, skin staple. This stapler comes with (35) wide medical grade stainless steel staples and measure 7.5mm in length, 4.3mm wide, and has a diameter of .6mm. This staper features: A unique ratchet system to prevent jamming Ergonomic, user friendly design Easy to check remaining staples in cartridge Clear view of operating site Short trigger stroke for ease of use and nonstop feed Angled head allows visibility of incision to ensure precise staple placement Alternative to products like ZipStitch instant stitches and Sutures Easy to use For deeper wounds Faster than Stitches Easier to remove than Stitches Fewer issues with scarring