Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Skinner List Semivolatiles, SPEX CertiPrep™
SDP

Catalog No. SKINNERSV Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SKINNER-SV 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SKINNER-SV Supplier SPEX CertiPrep Supplier No. SKINNERSV

missing translation for 'validMainframeMsg'

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,2-Dichlorobenzene95-50-12000μg/mL
1,3-Dichlorobenzene541-73-12000μg/mL
1,4-Dichlorobenzene106-46-72000μg/mL
1-Methylnaphthalene90-12-02000μg/mL
Anthracene120-12-72000μg/mL
Benz(a)anthracene56-55-32000μg/mL
Benzo(a)pyrene50-32-82000μg/mL
Benzo(b)fluoranthene205-99-22000μg/mL
Benzo(k)fluoranthene207-08-92000μg/mL
Bis(2-Ethylhexyl)phthalate117-81-72000μg/mL
Butylbenzyl phthalate85-68-72000μg/mL
Chrysene218-01-92000μg/mL
Diethyl phthalate84-66-22000μg/mL
Dimethyl phthalate131-11-32000μg/mL
Di-n-butyl phthalate84-74-22000μg/mL
Di-n-octyl phthalate117-84-02000μg/mL
Fluoranthene206-44-02000μg/mL
Indene95-13-62000μg/mL
Naphthalene91-20-32000μg/mL
Phenanthrene85-01-82000μg/mL
Pyrene129-00-02000μg/mL
Pyridine110-86-12000μg/mL
Quinoline91-22-52000μg/mL
7,12-Dimethylbenz(a)anthracene57-97-61000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Skinner List Semivolatiles
Packaging 2mL Amber Prescored Glass Ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solvent or Matrix MeCl
Quantity 1 mL

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.