Learn More
Fine Science Tools SKINNY BENT FORECEPS
SDP
Catalog No. NC9280124
Encompass
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 2.15 x 1.3mm/ Alloy / Material: Stainless Steel/ Length: 13cm
Catalog No. NC9280124 Supplier Fine Science Tools Supplier No. 1100313
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 2.15 x 1.3mm/ Alloy / Material: Stainless Steel/ Length: 13cm