Learn More
Medchemexpress LLC HY-101085 10mg Medchemexpress, SKL2001 CAS:909089-13-0 Purity:>98%
SDP
Catalog No. 501932030
Encompass_Preferred
Medchemexpress, HY-101085 10mg SKL2001 CAS:909089-13-0 SKL2001 is an agonist of the Wnt/β-catenin pathway, with anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2030 Supplier Medchemexpress LLC Supplier No. HY10108510MG
Medchemexpress, HY-101085 10mg SKL2001 CAS:909089-13-0 SKL2001 is an agonist of the Wnt/β-catenin pathway, with anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.