Learn More
Apexbio Technology LLC SKL2001 909089-13-0 200mg
SDP
Catalog No. 5000088049
Encompass_Preferred
SKL2001 (CAS 909089-13-0) is a small-molecule agonist that selectively activates the Wnt/ -catenin signaling pathway It functions by interfering with the interaction between Axin and -catenin thereby enhancing intracellular -catenin stability In cellular studies SKL2001 has been demonstrated to suppress cancer cell proliferation This compound is widely utilized in fundamental research investigating the molecular mechanisms of cancer and serves as a tool for drug discovery targeting the Wnt/ -catenin pathway
Catalog No. 50-000-88049 Supplier Apexbio Technology LLC Supplier No. B7920200
SKL2001 (CAS 909089-13-0) is a small-molecule agonist that selectively activates the Wnt/ -catenin signaling pathway It functions by interfering with the interaction between Axin and -catenin thereby enhancing intracellular -catenin stability In cellular studies SKL2001 has been demonstrated to suppress cancer cell proliferation This compound is widely utilized in fundamental research investigating the molecular mechanisms of cancer and serves as a tool for drug discovery targeting the Wnt/ -catenin pathway