Learn More
Sklar Instruments Freer Elevator, Sklar , 7-3/4 in., Sharp, Straight, Sharp/Blunt, Stainless Steel, Reusable. EA.
SDP
Catalog No. NC0940424 Shop All Sklar Instruments Products
Encompass_Preferred
Freer Elevator, Sklar, 7-3/4 in., Sharp, Straight, Sharp/Blunt, Stainless Steel, Reusable. Sklar's(R) Freer Elevator is an orthopedic instrument that can be used as a multipurpose tool in many procedures, although it is primarily used for blunt debulking and lifting periosteum from bones in confined areas. This double-ended instrument typically has a rounded handle with two slightly curved, tear-drop tips. One tip is blunt while the other is sharp. This product has 4.5mm tips and is 7-3/4 inches long. EA.
Catalog No. NC0940424 Supplier Sklar Instruments Supplier No. 472382
Freer Elevator, Sklar, 7-3/4 in., Sharp, Straight, Sharp/Blunt, Stainless Steel, Reusable. Sklar's(R) Freer Elevator is an orthopedic instrument that can be used as a multipurpose tool in many procedures, although it is primarily used for blunt debulking and lifting periosteum from bones in confined areas. This double-ended instrument typically has a rounded handle with two slightly curved, tear-drop tips. One tip is blunt while the other is sharp. This product has 4.5mm tips and is 7-3/4 inches long. EA.