Learn More
TARGETMOL CHEMICALS INC SKLB-163 5MG
SDP
Catalog No. 502580714
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SKLB-163 acts by downregulating RhoGDI activating JNK-1 signaling pathway and caspase-3 and reducing phosphorylated Akt and p44/42 MAPK. purity: 98%
Catalog No. 50-258-0714 Supplier TARGETMOL CHEMICALS INC Supplier No. T261935MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SKLB-163 acts by downregulating RhoGDI activating JNK-1 signaling pathway and caspase-3 and reducing phosphorylated Akt and p44/42 MAPK. purity: 98%