Learn More
Selleck Chemical LLC SKLB 610
SDP
Catalog No. 5000002657
Encompass_Preferred
SKLB-610 is a multi-target inhibitor of the tyrosine kinases It is most potent against VEGFR2 and exhibits slightly weaker inhibitor of FGFR2 and PDGFR
Catalog No. 50-000-02657 Supplier Selleck Chemical LLC Supplier No. S65265MG
SKLB-610 is a multi-target inhibitor of the tyrosine kinases It is most potent against VEGFR2 and exhibits slightly weaker inhibitor of FGFR2 and PDGFR