Learn More
TARGETMOL CHEMICALS INC SKLB 610 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T189510MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SKLB610 a novel multi-targeted inhibitor inhibits angiogenesis-related tyrosine kinase VEGFR2 FGFR2 and PDGFR at a rate of 97% 65% and 55% respectively at the concentration of 10 uM in biochemical kinase assays. purity: 99%
Catalog No. 50-257-7282