Learn More
TARGETMOL CHEMICALS INC SKLB1002 25MG
SDP
Catalog No. 502580822
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SKLB1002 is a potent and ATP-competitive VEGFR2 inhibitor with IC50 of 32 nM. purity: 99%
Catalog No. 50-258-0822 Supplier TARGETMOL CHEMICALS INC Supplier No. T242125MG
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SKLB1002 is a potent and ATP-competitive VEGFR2 inhibitor with IC50 of 32 nM. purity: 99%