Learn More
Medchemexpress LLC HY-139782 5mg , SKLB325 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1397825MG
Encompass_Preferred
HY-139782 5mg SKLB325 CAS: SKLB325 is a Jumonji domain-containing 6 (JMJD6) inhibitor with a binding affinity (KD) value of 0.755 μM, and the IC50 value of 0.7797 μM. SKLB325 exhibits antitumor effects on ovarian cancer in vivo and in vitro. SKLB325 induces apoptosis. SKLB325 exhibits remarkable antitumor efficacy in renal cell carcinoma (RCC) . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1451