Learn More
Selleck Chemical LLC SKLB4771 (FLT3-IN-1) S1099-2mg
SDP
Catalog No. 503111369
Encompass_Preferred
SKLB4771 is a potent and selective inhibitor of human receptor-type tyrosine-protein kinase FLT3 with IC50 of 10 nM
Catalog No. 50-311-1369 Supplier Selleck Chemical LLC Supplier No. S10992MG
SKLB4771 is a potent and selective inhibitor of human receptor-type tyrosine-protein kinase FLT3 with IC50 of 10 nM