Learn More
Lonza Walkersville HSMM – Human Skeletal Muscle Myoblasts
Supplier:  Lonza Walkersville CC2580
Encompass
Clonetics Human Skeletal Muscle Myoblasts (HSMM) are isolated from normal donors. HSMMs are isolated from the upper arm or leg muscle tissue and are sold in second passage. HSMMs can be differentiated to form myotubes in culture. Clonetics HSMMs are guaranteed through 10 population doublings and to differentiate to form multinucleated myotubes in culture. All cell types test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots. A Certificate of Analysis (CoA) is provided for each cell lot purchased.Applications: Cellular developmentDifferentiationDiabetesInsulin uptake or resistanceTissue repairDrug developmentMyotube developmentCo-culture with other cell typesContent:1 x Cryopreserved Ampule with ≥500,000 cells/vial
Catalog No. NC9074014