Learn More
ATCC SK-MEL-3; Malignant Melanoma; Human (Homo sapiens)
Catalog No. 502382764
Encompass
SK-MEL-3 is a cell line exhibiting fibroblast morphology that was isolated from the skin of a 42-year-old, White female patient with malignant melanoma. This product has applications in toxicology.
Catalog No. 50-238-2764 Supplier ATCC Supplier No. HTB69
SK-MEL-3 is a cell line exhibiting fibroblast morphology that was isolated from the skin of a 42-year-old, White female patient with malignant melanoma. This product has applications in toxicology.