Learn More
ATCC SK-MEL-5; Melanoma; Human (Homo sapiens)
Catalog No. 502384627
Encompass
SK-MEL-5 is a melanoma cell line isolated from skin tissue obtained from a 24-year-old, White, female malignant melanoma patient. This cell line is a suitable transfection host and also serves as a source of target cells for the detection of melanoma-specific antibody in patients with the disease. This is one of a very extensive series of melanoma lines isolated by T. Takahashi and associates.
Catalog No. 50-238-4627 Supplier ATCC Supplier No. HTB70
SK-MEL-5 is a melanoma cell line isolated from skin tissue obtained from a 24-year-old, White, female malignant melanoma patient. This cell line is a suitable transfection host and also serves as a source of target cells for the detection of melanoma-specific antibody in patients with the disease. This is one of a very extensive series of melanoma lines isolated by T. Takahashi and associates.