Learn More
ATCC SK-MES-1; Lung Carcinoma; Human (Homo sapiens)
Catalog No. 502383743
Encompass
SK-MES-1 is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 65-year-old, White male patient with squamous cell carcinoma. This product has applications in cancer research.
Catalog No. 50-238-3743 Supplier ATCC Supplier No. HTB58
 Restricted Item
Add to Cart
Add to Cart
SK-MES-1 is a cell line exhibiting epithelial morphology that was isolated from the lungs of a 65-year-old, White male patient with squamous cell carcinoma. This product has applications in cancer research.