Learn More
ATCC SK-N-BE(2); Neuroblastoma; Human (Homo sapiens)
Supplier:  ATCC CRL2271
Encompass
SK-N-BE(2) is a neuroblastoma cell line established in 1972 from a bone marrow biopsy taken from 2-year-old male with disseminated neuroblastoma after repeated courses of chemotherapy and radiotherapy. This cell line can be used in neuroscience and immunology research.
Catalog No. 50-238-3337