Learn More
ATCC SK-N-FI; Neuroblastoma; Human (Homo sapiens)
Catalog No. 502382100
Encompass
SK-N-FI are neuroblasts that were isolated from the brain of an 11-year-old, White, male patient with neuroblastoma. It has applications in immunology and neuroscience.
Catalog No. 50-238-2100 Supplier ATCC Supplier No. CRL2142
SK-N-FI are neuroblasts that were isolated from the brain of an 11-year-old, White, male patient with neuroblastoma. It has applications in immunology and neuroscience.