Learn More
MSC Skolnik Salvage Drum, 55 gal., UN Rated, 1 ea
Catalog No. 5000677815
Encompass
An open head salvage drum, constructed of high quality carbon steel, meets UN design type requirements. These standard salvage drums feature yellow bodies with yellow or red covers, and a multilingual salvage drum logo. Each steel salvage drum tests successfully on relevant international standards, including the 'T' rated salvage drum.
  • Constructed with 1.5/1.2/1.2 mm carbon steel
  • Dual UN ratings: 1A2/X400/S solid and 1A2/Y1.5/150 liquid
  • Bolt ring closure with EPDM gasket
  • Capacity: 55 gal.
  • Dimensions: 33 in. height, 22-1/2 in. diameter
  • Weight: 50 lb.
Catalog No. 50-006-77815 Supplier MSC Supplier No. 07572555
An open head salvage drum, constructed of high quality carbon steel, meets UN design type requirements. These standard salvage drums feature yellow bodies with yellow or red covers, and a multilingual salvage drum logo. Each steel salvage drum tests successfully on relevant international standards, including the 'T' rated salvage drum.
  • Constructed with 1.5/1.2/1.2 mm carbon steel
  • Dual UN ratings: 1A2/X400/S solid and 1A2/Y1.5/150 liquid
  • Bolt ring closure with EPDM gasket
  • Capacity: 55 gal.
  • Dimensions: 33 in. height, 22-1/2 in. diameter
  • Weight: 50 lb.