Learn More
Sigma Aldrich Fine Chemicals Biosciences SKOV3 Cells GFP STAT3
Catalog No. 502908520
Encompass
SKOV3 GFP-STAT3 are ovarian adenocarcinoma cells from a human 64 year old caucasion female having a ZFN modification creating a GFP-STAT3 transgene expressed from the endogenous STAT3 gene locus.This cell line was derived from ATCC Catalog No. HTB-77.
Catalog No. 50-290-8520 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLL11391VL
SKOV3 GFP-STAT3 are ovarian adenocarcinoma cells from a human 64 year old caucasion female having a ZFN modification creating a GFP-STAT3 transgene expressed from the endogenous STAT3 gene locus.This cell line was derived from ATCC Catalog No. HTB-77.