Learn More
TARGETMOL CHEMICALS INC SKP2 INHIBITOR C1 10MG
SDP
Catalog No. 502576785
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Skp2 Inhibitor C1 (SKPin C1)(SKPin C1) is a specific small molecule inhibitor of Skp2-mediated p27 degradation. purity: 99%
Catalog No. 50-257-6785 Supplier TARGETMOL CHEMICALS INC Supplier No. T425310MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Skp2 Inhibitor C1 (SKPin C1)(SKPin C1) is a specific small molecule inhibitor of Skp2-mediated p27 degradation. purity: 99%