Learn More
Selleck Chemical LLC Skp2 inhibitor C1 (SKPin C1) 5mg 432001-69-9
SDP
Supplier:  Selleck Chemical LLC S86525MG
Encompass_Preferred
Skp2 inhibitor C1 (SKPin C1) is a specific and selective small-molecule inhibitor of Skp2-mediated p27 degradation by reducing p27 binding through key compound-receptor contacts. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8294