Learn More
Cayman Chemical SKQ1bromIde 10mg
SDP
Catalog No. 502981464
Encompass_Preferred
A mitochondrial antioxidant; decreases transmembrane potential and production of ROS; prevents neuronal loss and synaptic damage in rat AD model; slows development of ocular pathologies; prevents acute phenoptosis following rhabdomyolysis, stroke, and myocardial infarction
Catalog No. 50-298-1464 Supplier Cayman Chemical Supplier No. 1989110 MG
A mitochondrial antioxidant; decreases transmembrane potential and production of ROS; prevents neuronal loss and synaptic damage in rat AD model; slows development of ocular pathologies; prevents acute phenoptosis following rhabdomyolysis, stroke, and myocardial infarction