Learn More
Cayman Chemical SKQ1 (bromide), Purity: ≥95%, CAS: 934826-68-3, 50 mg
SDP
Catalog No. 502985080
Encompass_Preferred
A mitochondrial antioxidant; decreases transmembrane potential and production of ROS; prevents neuronal loss and synaptic damage in rat AD model; slows development of ocular pathologies; prevents acute phenoptosis following rhabdomyolysis, stroke, and myocardial infarction
Catalog No. 50-298-5080 Supplier Cayman Chemical Supplier No. 1989150 MG
A mitochondrial antioxidant; decreases transmembrane potential and production of ROS; prevents neuronal loss and synaptic damage in rat AD model; slows development of ocular pathologies; prevents acute phenoptosis following rhabdomyolysis, stroke, and myocardial infarction