Learn More
Surgical Design™ Skull Crusher Hammer
Catalog No. 21124476 Shop All Surgical Design Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
21-124-476 Stainless Steel
1 options
Catalog No. 21-124-476 Supplier Surgical Design™ Supplier No. 1121101
Add to Cart
Add to Cart
Skull crusher hammer delivers controlled striking force for cranial opening, featuring solid stainless-steel construction and a knurled handle for secure grip and balance.
  • Constructed from medical-grade stainless steel for exceptional strength, balance, and corrosion resistance
  • Precision-weighted dual-face head provides uniform striking power and stability
  • Knurled handle grip ensures non-slip handling, even in gloved conditions
  • Designed for controlled impact in postmortem and osteological applications
  • One-piece forged construction enhances durability and reliability
  • Autoclavable and reusable, suitable for repeated laboratory or dissection use
  • Engineered for accuracy, balance, and user control in demanding work environments
  • Used in forensic, pathology, and postmortem examinations
  • Suitable for bone dissection, cranial access, and osteological studies
  • Common in medical training and anatomical research laboratories
Specifications
Product Type Skull Crusher
Material Stainless Steel
Sterility Non-sterile