Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Skullerz™ 3216 Microfiber Cleaning Cloth

Catalog No. 19820293 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-820-293 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-820-293 Supplier Ergodyne™ Supplier No. 19216
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Skullerz™ 3216 Microfiber Cleaning Cloth is designed to keep Skullerz™ specs clean and scratch-free.

  • Safely cleans lenses without scratching
  • Soft microfiber material
  • Embossed logo pattern

Specifications

Material Microfiber
Color Black
For Use With (Equipment) Skullerz™ safety glasses lenses
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.