Learn More
Ergodyne™ Skullerz™ 8945 Universal Bump Cap Insert
Catalog No. 19820754 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
6/Pk.
Catalog No. Quantity
19-820-754 6/Pk.
1 options
Catalog No. 19-820-754 Supplier Ergodyne™ Supplier No. 23380
Skullerz™ 8945 Universal Bump Cap Insert is a lightweight, impact-resistant shell that inserts into any cap, hat, or other headwear for additional head protection.
  • Horizontal and vertical two-way flex design adds protection from worker-generated impact to any cap
  • Innovative trim-to-fit feature adjusts height to fit different cap designs
  • Lightweight polypropylene construction will not break or crack
  • Strategically placed vents allow for continuous airflow and breathability
  • Closed cell foam padding reduces impact
  • Will not absorb odors
Specifications
Color Gray
Quantity 6/Pk.
Weight (English) < 3 oz.