Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Skullerz™ 8950 Bump Cap

Catalog No. 19820735 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Lime
Navy Blue
Size:
Long Brim
Micro Brim
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-820-735 Lime Micro Brim
19-820-736 - -
19-820-737 Lime Long Brim
19-820-740 - -
19-820-738 - -
19-820-742 - -
19-820-739 Navy Blue -
19-820-741 Navy Blue -
19-820-743 Navy Blue -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-820-735 Supplier Ergodyne™ Supplier No. 23331
Add to Cart
Add to Cart

Skullerz™ 8950 Bump Cap provides protection against bumps and bruises.

  • Removable impact-resistant shell with foam lining
  • Flanged design for comfort and air flow
  • Nylon or hi-vis polyester cap construction with reflective accents

Specifications

Size Micro Brim
Color Lime
Quantity 6/Pk.
For Use With (Application) Baggage Handlers, Manufacturing, Maintenance
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.