Learn More
Ergodyne™ Skullerz™ 8950XL Bump Cap
Catalog No. 19820747 Shop All Ergodyne Products
Encompass_Preferred
Catalog No. 19-820-747 Supplier Ergodyne™ Supplier No. 23349
Skullerz™ 8950XL Bump Cap provides protection against bumps and bruises.
  • Removable XL impact-resistant ABS plastic shell for larger heads and hairstyles
  • Recommended for hat size 7.5 or above
  • Flanged design for comfort and air flow
  • Nylon cap construction with reflective accents
  • EN 812-compliant