Learn More
Ergodyne™ Skullerz™ 8960 Bump Cap with LED Lighting Technology
Catalog No. 19820748 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Black
Lime Green
Navy Blue
Style:
Long Brim
Short Brim
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Style
19-820-748 Black Short Brim
19-820-750 Black Long Brim
19-820-752 Lime Green Short Brim
19-820-753 Lime Green Long Brim
19-820-749 Navy Blue Short Brim
19-820-751 Navy Blue Long Brim
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-820-748 Supplier Ergodyne™ Supplier No. 23370
Add to Cart
Add to Cart
Skullerz™ 8960 Bump Cap with LED Lighting Technology provides protection against bumps and bruises.
  • LED lighting technology built in under brim
  • LEDs emit 48 lumens; battery life of approximately 30 continuous hours
  • Powered by four CR2032 lithium ion batteries
  • Removable impact-resistant shell with foam lining
  • Flanged design for comfort and air flow
  • Nylon or hi-vis polyester with reflective accents
  • EN 812-compliant
Specifications
Color Black
Includes Under-brim LED lighting technology
Quantity 6/Pk.
Size Adjustable
Style Short Brim