Learn More
Ergodyne™ Skullerz™ Dagr Safety Glasses/Sunglasses, Kryptek™ Highlander™ Frame
Catalog No. 19910309 Shop All Ergodyne Products
Change view
Click to view available options
Lens Tint:
Clear
Clear with Fog-Off™
Copper
Polarized Copper
Polarized Smoke
Smoke
Smoke with Fog-Off™
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-910-309 Clear
19-910-310 Clear with Fog-Off™
19-910-312 Polarized Copper
19-910-313 Smoke
19-910-314 Polarized Smoke
19-910-315 Smoke with Fog-Off™
19-910-311 Copper
19-910-316 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-910-309 Supplier Ergodyne™ Supplier No. 52300
Add to Cart
Add to Cart
This item is not returnable. View return policy
Protective work safety sunglasses feature tough scratch-resistant lenses that filter 99.9% of harmful UVA, UVB and UVC rays to keep eyes safe in the sun and the blade-style design is a sleek and sporty look.
  • Durable half frames bend and flex without breaking
  • Non-slip rubber nose piece and rubber temples keep the glasses secure and resist sweat
  • Available with a polarized lens or with anti-fogging solution
  • Polarized glasses reduce glare and eye fatigue around water, snow and in other bright environments.
  • Anti-glare glasses block the harsh reflected light from water
  • Fog-Off™ treatment won't wash off or wear off for fog-free viewing and are ideal for use with dust masks
  • Certifications and testing:
    • Impact-resistant safety glasses are tested to ANSI Z87.1+ High-Velocity Impact Protection and optical requirements
    • Tested to MIL-PRF-32432 Ballistic Fragmentation, Class 1, which is the US Military Combat Eye Protection System Performance Specification for eyewear. To pass the military standard, safety glasses must withstand a steel projectile shot at 660 ft / 201m per second which is 4x faster than the Z87+ standard. Lenses must not puncture, crack or shatter. Frames must retain the lenses and not break.
  • Ideal for outdoor work like construction, carpentry/woodworking or landscaping or for outdoor activities like boating, skiing, fishing, hunting/shooting, and sports.
  • Available in a variety of lens options ideally suited for a variety of tasks and light environments
Specifications
Lens Coating Impact-resistant, Scratch-resistant, UV-treated
Certifications/Compliance ANSI Z87.1+, CSA Z94.3 + and MIL-PRF 32432 Ballistic Impact
Lens Tint Clear
For Use With (Application) Construction/Trades, Oil and Gas, Mining, Maintenance, Assembly/Fabrication, Manufacturing, Police/Tactical
No. per Case 12/Cs.