Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Skullerz™ Elastic Retainer Strap

Catalog No. 19910429 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-910-429 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-910-429 Supplier Ergodyne™ Supplier No. 59991
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Skullerz™ Elastic Retainer Strap features a locking system that secures elastic straps to temples.

  • Inserts into temple slots, twists, and locks
  • Use alone or with foam gasket insert
  • For use with Odin, Dagr, Kvasir, or Valkyrie Safety Glasses

Specifications

Color Black
For Use With (Equipment) Skullerz™ Safety Glasses
For Use With (Application) Creating a snug and secure fit
No. per Pack 12/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.