Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Skullerz™ Kvasir Foam Gasket Insert

Catalog No. 19910349 Shop All Ergodyne Products
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-910-349 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-910-349 Supplier Ergodyne™ Supplier No. 53991

Skullerz™ Kvasir Foam Gasket Insert is a removable insert that keep out dust and debris.

  • Reduces dust and debris entry
  • Snaps in place at corners
  • Easily removes when not desired or for cleaning
  • Use alone or with elastic retainer strap
  • Fits Kvasir safety glasses only

Specifications

Color Black
For Use With (Equipment) Kvasir Safety Glasses
For Use With (Application) Reducing dust and debris entry
No. per Pack 12/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.