Learn More
Ergodyne™ Skullerz™ Sköll Safety Glasses/Sunglasses
Catalog No. 19910428 Shop All Ergodyne Products
Encompass_Preferred
Change view
Click to view available options
Frame Color:
Black
Lens Tint:
Clear
Clear with Fog-Off™
Indoor/Outdoor
Indoor/Outdoor with Fog-Off™
Smoke
Smoke with Fog-Off™
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-910-428 Black Indoor/Outdoor with Fog-Off™
19-910-423 Black Clear
19-910-424 Black Clear with Fog-Off™
19-910-425 Black Smoke
19-910-426 Black Smoke with Fog-Off™
19-910-427 Black Indoor/Outdoor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-910-428 Supplier Ergodyne™ Supplier No. 59083
This item is not returnable. View return policy
Potective work safety sunglasses feature tough, scratch-resistant lenses filter 99.9% of harmful UVA, UVB and UVC rays to keep eyes safe in the sun.
  • Blade-style design is a sleek and sporty look
  • Durable frames bend and flex without breaking
  • Flexible rubber temples and non-slip nose piece offer a non-slip, comfortable fit
  • Anti-glare glasses block the harsh reflected light from water, keeping eyes comfortable around glare.
  • Fog-Off™ treatment won't wash off or wear off and is ideal for use with dust masks.
  • Certifications and Testing:
    • Impact-resistant safety glasses are tested to ANSI Z87.1+ High-Velocity Impact Protection and optical requirements
    • Tested to MIL-PRF-32432 Ballistic Fragmentation, Class 1, which is the US Military Combat Eye Protection System Performance Specification for eyewear. To pass the military standard, safety glasses must withstand a steel projectile shot at 660 ft / 201m per second which is 4x faster than the Z87+ standard. Lenses must not puncture, crack or shatter. Frames must retain the lenses and not break.
  • Ideal for protecting eyes during outdoor work like construction, carpentry/woodworking or landscaping or for outdoor activities like boating, skiing, fishing, hunting/shooting, and sports.
  • Available in a variety of lens options ideally suited for a variety of tasks and light environments.
Specifications
Lens Coating Impact-resistant, Scratch-resistant, UV-treated
Certifications/Compliance ANSI Z87.1+, CSA Z94.3 and MIL-PRF 32432 Ballistic Impact
Lens Tint Indoor/Outdoor with Fog-Off™
For Use With (Application) Construction/Trades,Oil and Gas,Mining,Maintenance,Assembly/Fabrication,Manufacturing,Police/Tactical
Frame Color Black
No. per Case 12/Cs.