Learn More
MSC Ergodyne Face Shield, Skullerz 8989, Mesh, Hard Hat Compatible, 1 ea
Catalog No. 5000685061
Encompass
This face shield is designed for integration with hard hats and safety helmets to provide increased eye and face protection. Featuring durable PC-ABS material and metal mesh construction, this shield offers full coverage and protects against flying debris while maintaining high breathability and preventing fogging. It is suitable for tasks such as forestry, landscaping, and environments with airborne debris.
  • Meets ANSI/ISEA Z87.1-2020 impact resistance standards
  • Includes adapter for mounting into side accessory slots
  • Pivots into three positions: fully down, mid-way, and fully up
  • Soft visor conforms to the helmet shell to block dust
  • Compatible with accessories like headlamps and earmuffs
  • Adapter pivots forward and backward for a customized fit
  • Removable mesh shield design
Catalog No. 50-006-85061 Supplier MSC Supplier No. 73659518
This face shield is designed for integration with hard hats and safety helmets to provide increased eye and face protection. Featuring durable PC-ABS material and metal mesh construction, this shield offers full coverage and protects against flying debris while maintaining high breathability and preventing fogging. It is suitable for tasks such as forestry, landscaping, and environments with airborne debris.
  • Meets ANSI/ISEA Z87.1-2020 impact resistance standards
  • Includes adapter for mounting into side accessory slots
  • Pivots into three positions: fully down, mid-way, and fully up
  • Soft visor conforms to the helmet shell to block dust
  • Compatible with accessories like headlamps and earmuffs
  • Adapter pivots forward and backward for a customized fit
  • Removable mesh shield design