Learn More
Andwin Scientific SKURA CUTTING BLADE PK/5
SDP
Catalog No. NC9281730
Encompass_Preferred
The Coverslipping Film Cutter Blades are accessories for the Tissue-Tek Film Coverslippers, a ready-to-use product that offers superior performance, convenience, and flexibility in staining and glass coverslipping. This product by Sakura features a quick action of coverslipping 360 slides per hour. It is sold separately. The blades are sold in a pack of 5.
Catalog No. NC9281730 Supplier Andwin Scientific Supplier No. 4772
The Coverslipping Film Cutter Blades are accessories for the Tissue-Tek Film Coverslippers, a ready-to-use product that offers superior performance, convenience, and flexibility in staining and glass coverslipping. This product by Sakura features a quick action of coverslipping 360 slides per hour. It is sold separately. The blades are sold in a pack of 5.