Learn More
MSC Sky Hook Lifting Crane, 500 lb Capacity, Receiver Hitch Mount, Chain Drive
Catalog No. 5000533183
Encompass
This portable lifting crane mounts into a standard 2 inch receiver hitch, providing an efficient material handling solution. Built from high-strength steel, it safely lifts and positions loads up to 500 lb. Its chain drive and friction brake offer precise control, ideal for applications requiring portability and safe lifting.
  • Prevents lifting injuries.
  • Requires no drilling or welding for installation.
  • Lightweight and portable with removable boom for storage.
  • Enables one person to safely lift and position loads up to 500 lb.
  • Designed for precision positioning in tight working conditions.
  • Features an economical lever-operated friction brake for controlled operation.
  • Constructed of high-strength steel.
  • Meets applicable OSHA and ASME requirements.
  • Made in the U.S.A.
Catalog No. 50-005-33183 Supplier MSC Supplier No. 08635096
This portable lifting crane mounts into a standard 2 inch receiver hitch, providing an efficient material handling solution. Built from high-strength steel, it safely lifts and positions loads up to 500 lb. Its chain drive and friction brake offer precise control, ideal for applications requiring portability and safe lifting.
  • Prevents lifting injuries.
  • Requires no drilling or welding for installation.
  • Lightweight and portable with removable boom for storage.
  • Enables one person to safely lift and position loads up to 500 lb.
  • Designed for precision positioning in tight working conditions.
  • Features an economical lever-operated friction brake for controlled operation.
  • Constructed of high-strength steel.
  • Meets applicable OSHA and ASME requirements.
  • Made in the U.S.A.