Learn More
AdipoGen Skyrin (1 mg)
SDP
Supplier:  AdipoGen AGCN20001M001
Encompass_Preferred
Chemical. CAS: 602-06-2. Formula: C30H18O10. MW: 538.5. Non-peptidic anti-diabetic agent. Receptor-selective glucagon antagonist. Free radical species (·OH, R·) and singlet oxygen (1O2) scavenger. Mycotoxin. Cytotoxic. Antioxidant. Inhibitor of botulinum neurotoxin serotype A (BoNTA). Anti-MRSA compound. Antibiotic.
Catalog No. 50-205-3350