Learn More
MSC SkyScan Lightning Detector, 40 Mile Range, Handheld
Catalog No. 5000533048
Encompass
This hand-held lightning detector is designed for weather monitoring, detecting lightning and thunderstorm activity within 40 miles. It uses patented technology with a full digital microprocessor for accuracy and reliability.
  • Extended operating life with 6 AA batteries (not included).
  • Detects lightning and thunderstorm activity up to 40 miles away.
  • Patented technology determines distance to detected strokes.
  • Indicates distances in four ranges: 0-3, 3-8, 8-20, and 20-40 miles.
  • Determines if storm is moving towards, away, or parallel to your position.
  • Emits an audible warning tone upon lightning detection.
  • Range indicator lights up for 3 seconds, showing distance to last stroke.
  • Automatic shut-off after five hours of continuous operation for extended battery life.
Catalog No. 50-005-33048 Supplier MSC Supplier No. 09175639
This hand-held lightning detector is designed for weather monitoring, detecting lightning and thunderstorm activity within 40 miles. It uses patented technology with a full digital microprocessor for accuracy and reliability.
  • Extended operating life with 6 AA batteries (not included).
  • Detects lightning and thunderstorm activity up to 40 miles away.
  • Patented technology determines distance to detected strokes.
  • Indicates distances in four ranges: 0-3, 3-8, 8-20, and 20-40 miles.
  • Determines if storm is moving towards, away, or parallel to your position.
  • Emits an audible warning tone upon lightning detection.
  • Range indicator lights up for 3 seconds, showing distance to last stroke.
  • Automatic shut-off after five hours of continuous operation for extended battery life.