Learn More
Medchemexpress LLC HY-15237 1mg Medchemexpress, SL 0101-1 CAS:77307-50-7 Purity:>98%
SDP
Catalog No. 501932728
Encompass_Preferred
Medchemexpress, HY-15237 1mg SL 0101-1 CAS:77307-50-7 SL 0101-1 (SL0101), a kaempferol glycoside, isolated from the tropical plant F. refracta, is a cell-permeable, selective, reversible, ATP-competitive p90 Ribosomal S6 Kinase (RSK) inhibitor, with an IC 50 of 89 nM. Shows proliferation inhibition in human breast cancer cell line MCF-7 and produces a cell cycle block in G 1 phase [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2728 Supplier Medchemexpress LLC Supplier No. HY152371MG
Medchemexpress, HY-15237 1mg SL 0101-1 CAS:77307-50-7 SL 0101-1 (SL0101), a kaempferol glycoside, isolated from the tropical plant F. refracta, is a cell-permeable, selective, reversible, ATP-competitive p90 Ribosomal S6 Kinase (RSK) inhibitor, with an IC 50 of 89 nM. Shows proliferation inhibition in human breast cancer cell line MCF-7 and produces a cell cycle block in G 1 phase [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.