Learn More
Enzo Life Sciences SL-327 (10mg). CAS: 305350-87-2
SDP
Supplier:  Enzo Life Sciences BMLEI3650010
Encompass_Preferred
A selective inhibitor of MEK1 and MEK2 (IC50= 0.18 and 0.22 µM respectively). Displays neuroprotective effects against ischemic brain injury. Alternative name: [Amino[(4-aminophenyl)thio]methylene]-2-(trifluoromethyl)benzeneacetonitrile. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO to 100 mM or in ethanol to 100 mM. Long Term Storage: -20°C.
Catalog No. 50-201-0748