Learn More
Selleck Chemical LLC SL-327-25mg
SDP
Catalog No. 502389873
Encompass_Preferred
SL327 is a selective inhibitor for MEK1/2 with IC50 of 0.18 μM/ 0.22 μM, no activity towards Erk1, MKK3, MKK4, c-JUN, PKC, PKA, or CamKII;capable of transport through the blood-brain barrier.
Catalog No. 50-238-9873 Supplier Selleck Chemical LLC Supplier No. S106625MG
SL327 is a selective inhibitor for MEK1/2 with IC50 of 0.18 μM/ 0.22 μM, no activity towards Erk1, MKK3, MKK4, c-JUN, PKC, PKA, or CamKII;capable of transport through the blood-brain barrier.