Learn More
Apexbio Technology LLC SL-327, 5mg. Cas: 305350-87-2 MFCD: MFCD06411432. Selective MEK1/2 inhibitor
SDP
Catalog No. 501904718
Encompass_Preferred
SL-327 is a selective inhibitor of MEK1 and MEK2 with IC50 values of 0.18 and 0.22M, respectively.MEK1 and MEK2 (ERK) are a kinase enzyme which phosphorylate mitogen-activated protein kinase (MAPK). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4718 Supplier Apexbio Technology LLC Supplier No. A18945
SL-327 is a selective inhibitor of MEK1 and MEK2 with IC50 values of 0.18 and 0.22M, respectively.MEK1 and MEK2 (ERK) are a kinase enzyme which phosphorylate mitogen-activated protein kinase (MAPK). Other sizes are available. Please inqury us for quote.