Learn More
Pyramex™ SL Series Cap Style Hard Hat
Catalog No. 19155406 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Product Type:
4-Point Ratchet
4-Point Snap Lock
6-Point Ratchet
6-Point Snap Lock
Color:
Black
Blue
Dark Blue
Green
Hi-vis Lime
Hi-vis Orange
Hi-vis Pink
Light Blue
Orange
Red
White
Yellow
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color
19-155-406 6-Point Ratchet Blue
19-155-372 4-Point Snap Lock Red
19-155-373 4-Point Snap Lock Yellow
19-155-374 4-Point Snap Lock Hi-vis Lime
19-155-375 4-Point Snap Lock Green
19-155-376 4-Point Snap Lock Orange
19-155-377 4-Point Snap Lock Blue
19-155-381 4-Point Ratchet Black
19-155-385 4-Point Ratchet Hi-vis Lime
19-155-387 4-Point Ratchet Orange
19-155-388 4-Point Ratchet Hi-vis Orange
19-155-390 4-Point Ratchet Light Blue
19-155-391 4-Point Ratchet Dark Blue
19-155-392 4-Point Ratchet Hi-vis Pink
19-155-393 6-Point Snap Lock White
19-155-394 6-Point Snap Lock Red
19-155-395 6-Point Snap Lock Yellow
19-155-397 6-Point Snap Lock Orange
19-155-400 6-Point Ratchet White
19-155-401 6-Point Ratchet Red
19-155-402 6-Point Ratchet Yellow
19-155-403 6-Point Ratchet Green
19-155-404 6-Point Ratchet Orange
19-155-405 6-Point Ratchet Hi-vis Orange
19-151-528 4-Point Snap Lock White
19-155-378 4-Point Snap Lock Light Blue
19-155-396 6-Point Snap Lock Green
19-155-398 6-Point Snap Lock Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 options
Catalog No. 19-155-406 Supplier Pyramex™ Supplier No. HP16160
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Exceeds ANSI Z89.1, Type 1, Class C, G, and E for industrial head protection; CSA Z94.1-15
  • Low profile design
  • Tested and marked for reverse donning
  • Shell constructed from High Density Polyethylene materials
  • Rain trough on sides and back of helmet channel moisture away
  • Soft brow pad is replaceable. Replaceable suspensions and headbands also available
  • Snap Lock suspension features 'tuck away' adjustments for all day, hassle free wear
  • Ratchet suspension is easy to adjust and allows the wearer to modify the fit while wearing hard hat
  • Universal accessory slots fit cap mounted ear muffs and accessories
  • Hats also accommodate chin straps
  • Testing: Impact, Penetration, Flammability, Electrical Insulation, Marking and Humidity
  • Specifications
    Certifications/Compliance ANSI Z89.1 standards, Type 1 --- Class C, G, and E
    Color Blue
    Material HDPE
    Product Type 6-Point Ratchet