Learn More
Pyramex Safety Products SL Series Sleek Shell Hard Hat
Catalog No. 19555119 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Head Style:
Front Brim
Full Brim
Color:
Blue
Gray
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Head Style Color
19-555-119 Full Brim Gray
19-555-114 Front Brim White
19-555-118 Full Brim White
19-555-115 Front Brim Gray
19-555-116 Front Brim Red
19-555-117 Front Brim Blue
19-555-120 Full Brim Red
19-555-121 Full Brim Yellow
19-555-122 Full Brim Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-555-119 Supplier Pyramex Safety Products Supplier No. HPS24112
Add to Cart
Add to Cart
Pyramex Safety™ SL Series Sleek Shell Hard Hat offers impact and penetration resistance.
  • Ridgeless low profile design
  • No rain trough-prevents pooling of liquids
  • Soft brow pad absorbs perspiration
Specifications
Color Gray
Material Polyethylene
Suspension Type 4-point ratchet suspension
Head Style Full Brim