Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing, 2-Stop, Tygon LMT-55, 1.85 mm ID; 12/PK
Catalog No. NC1592994
Encompass
The overall length of LMT-55 97618-series stop tubing is 16" (406 mm). The legacy Ismatec stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
Catalog No. NC1592994 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9761840
The overall length of LMT-55 97618-series stop tubing is 16" (406 mm). The legacy Ismatec stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.