Learn More
Plastilite Corp SL22 SHIPPING COOLER
Catalog No. NC0284997
Encompass_Preferred
This item has a minimum order quantity of 7 per supplier requirements.
Mailing and Shipping Containers, Insulated Containers. EPS Shipper. Foam Insulated Container. Foam Shipper. Insulated Shipper. Insulated Shipper Container. Temperature-controlled Package Shipper. 1.5” thick-wall foam cooler with white outer box at 32ECT strength with “PERISHABLE” and up arrows on two sides. Durable and highly insulating for your temperature sensitive products such as pharmaceutical and biological materials. Use with dry ice, gel ice packs, foam brick ice packs, and PCMs as refrigerant material. With proper amounts of refrigerant material this cooler can protect up to four days depending on ambient shipping temperatures. SL22-C cooler in box, 1.5” thick. Inside Dimensions of 16”x12”x12.625” offering 1.4 cubic feet for product and refrigerant material. This EPS foam cooler is recyclable at more than 400 locations across North America.
Catalog No. NC0284997 Supplier Plastilite Corp Supplier No. SL22C
This item has a minimum order quantity of 7 per supplier requirements.
Mailing and Shipping Containers, Insulated Containers. EPS Shipper. Foam Insulated Container. Foam Shipper. Insulated Shipper. Insulated Shipper Container. Temperature-controlled Package Shipper. 1.5” thick-wall foam cooler with white outer box at 32ECT strength with “PERISHABLE” and up arrows on two sides. Durable and highly insulating for your temperature sensitive products such as pharmaceutical and biological materials. Use with dry ice, gel ice packs, foam brick ice packs, and PCMs as refrigerant material. With proper amounts of refrigerant material this cooler can protect up to four days depending on ambient shipping temperatures. SL22-C cooler in box, 1.5” thick. Inside Dimensions of 16”x12”x12.625” offering 1.4 cubic feet for product and refrigerant material. This EPS foam cooler is recyclable at more than 400 locations across North America.