Learn More
Cole-Parmer AEMC SL261 AC/DC Current Probe, 100mA to 100A, Lead w/ BNC
Supplier:  Cole-Parmer 2000215
Encompass
This clamp-on current proble ranges 100mA to 3000Amrs and measures AC, DC and AC/DC. It should be used with DMMs, oscilloscopes, dataloggers, handheld scopes and power meters. It has a voltage or current output.
Catalog No. 50-992-591