Learn More
International Crystal Model SL-3 NaCl Sealed Liquid Cell with rectangular (dispersive) aperture
Catalog No. NC1362442
Encompass_Preferred
The Model SL-3 stainless steel sealed liquid cell has been designed to permit accurate alignment of the cell components. Alignment is accomplished by placing the front plate face down on a block of wood or other object about 3/4' thick and then placing the front lead gasket with holes in alignment with the hypodermic ports. The front drilled window is then placed in position over the ports, which are still visible. The amalgamated spacer, rear window, neoprene gasket and back plate can then be positioned without difficulty to seal the cell. The front gasket and front plate of the SL-3 are amalgamated permanently. The aperture of the dispersive cell is 22.5mm x 10mm. The window size is 38.5mmx19.5mmx4mm.
Catalog No. NC1362442 Supplier International Crystal Supplier No. 0005A590W
The Model SL-3 stainless steel sealed liquid cell has been designed to permit accurate alignment of the cell components. Alignment is accomplished by placing the front plate face down on a block of wood or other object about 3/4' thick and then placing the front lead gasket with holes in alignment with the hypodermic ports. The front drilled window is then placed in position over the ports, which are still visible. The amalgamated spacer, rear window, neoprene gasket and back plate can then be positioned without difficulty to seal the cell. The front gasket and front plate of the SL-3 are amalgamated permanently. The aperture of the dispersive cell is 22.5mm x 10mm. The window size is 38.5mmx19.5mmx4mm.